Assay Details
| Gene Symbol: UBE2D2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014730637.2 | XP_014586123.1 | 306 | 101 | ||
| XM_023617596.1 | XP_023473364.1 | 729 | 101 | |||
More Information
Set Membership: |
|

UBE2D2
Genomic Map