Assay Details
| Gene Symbol: ST13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001309324.1 | NP_001296253.1 | 5 - 6 | 385 | 93 | |
| GenBank mRNA | JL616190.1 | - | 5 - 6 | 391 | 93 | |
More Information
Set Membership: |
|

ST13
Genomic Map