Assay Details
| Gene Symbol: HAS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081801.2 | NP_001075270.1 | 3 - 4 | 1158 | 78 | |
| GenBank mRNA | AY056582.1 | - | 3 - 4 | 1161 | 78 | |
More Information
Set Membership: |
|

HAS2
Genomic Map