Assay Details
| Gene Symbol: LY49F | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081830.1 | NP_001075299.1 | 7 - 8 | 931 | 134 | |
| XM_005610809.3 | XP_005610866.1 | 868 | 134 | |||
| GenBank mRNA | AB120393.1 | - | 7 - 8 | 931 | 134 | |
More Information
Set Membership: |
|

LY49F
Genomic Map