Assay Details
| Gene Symbol: OPN1LW | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081845.1 | NP_001075314.1 | 4 - 5 | 759 | 71 | |
| GenBank mRNA | AF031531.1 | - | 3 - 4 | 604 | 71 | |
| AF132043.1 | - | 4 - 5 | 759 | 71 | ||
More Information
Set Membership: |
|

OPN1LW
Genomic Map