Assay Details
| Gene Symbol: NR5A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081851.1 | NP_001075320.1 | 5 - 6 | 1150 | 98 | |
| GenBank mRNA | AF203911.1 | - | 5 - 6 | 1150 | 98 | |
More Information
Set Membership: |
|

NR5A1
Genomic Map