Assay Details
| Gene Symbol: CCL11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081871.1 | NP_001075340.1 | 1 - 2 | 144 | 146 | |
| GenBank mRNA | AJ251188.1 | - | 1 - 2 | 144 | 146 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AJ251188.1, NM_001081871.1 transcripts.
Set Membership: |
|

CCL11
Genomic Map