Assay Details
| Gene Symbol: S100A7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081880.1 | NP_001075349.1 | 1 - 2 | 190 | 128 | |
| XM_014739696.2 | XP_014595182.1 | 267 | 128 | |||
| GenBank mRNA | AJ781506.1 | - | 1 - 2 | 190 | 128 | |
More Information
Set Membership: |
|

S100A7
Genomic Map