Assay Details
| Gene Symbol: DEFB4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081887.1 | NP_001075356.1 | 1 - 2 | 88 | 81 | |
| GenBank mRNA | AY170305.1 | - | 1 - 2 | 88 | 81 | |
More Information
Set Membership: |
|

DEFB4B
Genomic Map