Assay Details
| Gene Symbol: ANG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081899.1 | NP_001075368.1 | 1 | 352 | 96 | |
| XM_005602593.3 | XP_005602650.1 | 505 | 96 | |||
| XM_014733444.2 | XP_014588930.1 | 441 | 96 | |||
| GenBank mRNA | AY450362.1 | - | 1 | 352 | 96 | |
More Information
Set Membership: |
|

ANG
Genomic Map