Assay Details
| Gene Symbol: FABP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081903.2 | NP_001075372.1 | 1 - 2 | 103 | 77 | |
| GenBank mRNA | AY536518.1 | - | 1 - 2 | 104 | 77 | |
More Information
Set Membership: |
|

FABP2
Genomic Map