Assay Details
| Gene Symbol: AKR1C1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081908.1 | NP_001075377.1 | 10 - 11 | 1271 | 99 | |
| XM_023631879.1 | XP_023487647.1 | 1190 | 99 | |||
| GenBank mRNA | AY955082.1 | - | 10 - 11 | 1271 | 99 | |
More Information
Set Membership: |
|

AKR1C1
Genomic Map