Assay Details
| Gene Symbol: LY49B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081923.1 | NP_001075392.1 | 4 - 5 | 347 | 107 | |
| GenBank mRNA | AB120380.1 | - | 4 - 5 | 347 | 107 | |
| AB120381.1 | - | 3 - 4 | 248 | 107 | ||
| AB120382.1 | - | 3 - 4 | 310 | 107 | ||
| AB120383.1 | - | 3 - 4 | 310 | 107 | ||
| AB120384.2 | - | 3 - 4 | 310 | 107 | ||
More Information
Set Membership: |
|

LY49B
Genomic Map