Assay Details
| Gene Symbol: LY49D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081924.1 | NP_001075393.1 | 5 - 6 | 642 | 149 | |
| GenBank mRNA | AB120387.1 | - | 5 - 6 | 642 | 149 | |
| AB120388.1 | - | 5 - 6 | 624 | 149 | ||
More Information
Set Membership: |
|

LY49D
Genomic Map