Assay Details
| Gene Symbol: SERPINB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081947.1 | NP_001075416.1 | 3 - 4 | 349 | 113 | |
| XM_005603498.3 | XP_005603555.1 | 752 | 113 | |||
| XM_005603499.3 | XP_005603556.1 | 342 | 113 | |||
| XM_005603500.3 | XP_005603557.1 | 625 | 113 | |||
| GenBank mRNA | JL616060.2 | - | 2 - 3 | 253 | 113 | |
| M91161.1 | - | 3 - 4 | 349 | 113 | ||
More Information
Set Membership: |
|

SERPINB1
Genomic Map