Assay Details
| Gene Symbol: HAPLN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082504.1 | NP_001075973.1 | 4 - 5 | 1116 | 118 | |
| GenBank mRNA | X78077.1 | - | 1116 | 118 | ||
More Information
Set Membership: |
|

HAPLN1
Genomic Map