Assay Details
| Gene Symbol: CIAO2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099442.1 | NP_001092912.1 | 4 - 5 | 395 | 65 | |
| XM_005602597.3 | XP_005602654.2 | 519 | 65 | |||
| GenBank mRNA | EF397504.1 | - | 4 - 5 | 395 | 65 | |
More Information
Set Membership: |
|

CIAO2A
Genomic Map