Assay Details
| Gene Symbol: CYP2A13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111337.1 | NP_001104807.1 | 1 - 2 | 182 | 70 | |
| GenBank mRNA | EU286274.1 | - | 1 - 2 | 182 | 70 | |
More Information
Set Membership: |
|

CYP2A13
Genomic Map