Assay Details
Gene Symbol: CAV2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114144.1 | NP_001107616.1 | 2 - 3 | 820 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
