Assay Details
| Gene Symbol: CXCL10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114940.1 | NP_001108412.1 | 2 - 3 | 186 | 146 | |
| GenBank mRNA | EU438777.1 | - | 2 - 3 | 186 | 146 | |
| HQ890261.1 | - | 2 - 3 | 252 | 146 | ||
More Information
Set Membership: |
|

CXCL10
Genomic Map