Assay Details
| Gene Symbol: PTGER2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127352.1 | NP_001120824.1 | 1 - 2 | 838 | 80 | |
| GenBank mRNA | EF569555.1 | - | 1 - 2 | 838 | 80 | |
| Gene Symbol: LOC111769596 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023625271.1 | XP_023481039.1 | 1332 | 80 | ||
More Information
Set Membership: |
|

PTGER2
Genomic Map