Assay Details
| Gene Symbol: CXCL9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130078.1 | NP_001123550.1 | 2 - 3 | 185 | 89 | |
| XM_023636871.1 | XP_023492639.1 | 496 | 89 | |||
| GenBank mRNA | EU438776.1 | - | 2 - 3 | 185 | 89 | |
More Information
Set Membership: |
|

CXCL9
Genomic Map