Assay Details
| Gene Symbol: HMGB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081835.2 | NP_001075304.1 | 5 | 1066 | 156 | |
| GenBank mRNA | AB275457.1 | - | 5 | 1056 | 156 | |
| JL616373.1 | - | 5 | 1122 | 155 | ||
More Information
Set Membership: |
|

HMGB1
Genomic Map