Assay Details
| Gene Symbol: CCL8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081864.2 | NP_001075333.2 | 3 | 678 | 162 | |
| GenBank mRNA | AF506972.1 | - | 3 | 652 | 163 | |
| BM734601.1 | - | 2 | 491 | 162 | ||
More Information
Set Membership: |
|

CCL8
Genomic Map