Assay Details
| Gene Symbol: HSP90AA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163955.1 | NP_001157427.1 | 9 - 10 | 2087 | 109 | |
| XM_023627692.1 | XP_023483460.1 | 2265 | 109 | |||
| GenBank mRNA | AB043677.1 | - | 9 - 10 | 2054 | 109 | |
| JL617688.1 | - | 7 - 8 | 1539 | 109 | ||
More Information
Set Membership: |
|

HSP90AA1
Genomic Map