Assay Details
| Gene Symbol: A4GNT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163877.1 | NP_001157349.1 | 1 - 2 | 414 | 78 | |
| XM_023619735.1 | XP_023475503.1 | 535 | 78 | |||
| GenBank mRNA | AB256017.1 | - | 1 - 2 | 221 | 78 | |
More Information
Set Membership: |
|

A4GNT
Genomic Map