Assay Details
| Gene Symbol: SOX9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023652130.1 | XP_023507898.1 | 982 | 93 | ||
| GenBank mRNA | AF265207.1 | - | 1 | 150 | 93 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: SOX9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023652130.1 | XP_023507898.1 | 982 | 93 | ||
| GenBank mRNA | AF265207.1 | - | 1 | 150 | 93 | |
Set Membership: |
|