Assay Details
| Gene Symbol: LOC100033941 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005607588.3 | XP_005607645.1 | 1566 | 88 | ||
| XM_005607589.3 | XP_005607646.2 | 1929 | 88 | |||
| XM_023635991.1 | XP_023491759.1 | 1970 | 88 | |||
| XM_023635992.1 | XP_023491760.1 | 1964 | 88 | |||
| GenBank mRNA | AF547431.1 | - | 2 - 3 | 205 | 88 | |
More Information
Set Membership: |
|

LOC100033941
Genomic Map