Assay Details
| Gene Symbol: CCL13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163887.1 | NP_001157359.1 | 1 - 2 | 78 | 116 | |
| GenBank mRNA | AJ251191.1 | - | 1 - 2 | 63 | 116 | |
More Information
Set Membership: |
|

CCL13
Genomic Map