Assay Details
| Gene Symbol: RACK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001242446.1 | NP_001229375.1 | 4 - 5 | 634 | 58 | |
| GenBank mRNA | AY246708.1 | - | 2 - 3 | 153 | 58 | |
| CD464593.1 | - | 2 - 3 | 159 | 58 | ||
| CD466381.1 | - | 4 - 5 | 634 | 58 | ||
| JL639002.1 | - | 2 - 3 | 248 | 58 | ||
More Information
Set Membership: |
|

RACK1
Genomic Map