Assay Details
| Gene Symbol: SIAH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014731540.2 | XP_014587026.2 | 520 | 85 | ||
| GenBank mRNA | AY246715.1 | - | 1 - 2 | 204 | 85 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SIAH2| Gene Symbol: SIAH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014731540.2 | XP_014587026.2 | 520 | 85 | ||
| GenBank mRNA | AY246715.1 | - | 1 - 2 | 204 | 85 | |
Set Membership: |
|