Assay Details
| Gene Symbol: RPS12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001309481.1 | NP_001296410.1 | 3 - 4 | 218 | 66 | |
| GenBank mRNA | AY246731.1 | - | 1 - 2 | 109 | 66 | |
| CX593239.1 | - | 3 - 4 | 241 | 66 | ||
More Information
Set Membership: |
|

RPS12
Genomic Map