Assay Details
| Gene Symbol: FILIP1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163896.1 | NP_001157368.1 | 5 | 3173 | 81 | |
| XM_005601759.3 | XP_005601816.1 | 3240 | 81 | |||
| XM_005601762.3 | XP_005601819.1 | 2249 | 81 | |||
| XM_023623129.1 | XP_023478897.1 | 2834 | 81 | |||
| GenBank mRNA | AY349169.1 | - | 1 | 250 | 81 | |
More Information
Set Membership: |
|

FILIP1L
Genomic Map