Assay Details
Gene Symbol: IGFBP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001308603.2 | NP_001295532.1 | 1 - 2 | 1090 | 62 | |
GenBank mRNA | AY752934.1 | - | 1 - 2 | 214 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
