Assay Details
| Gene Symbol: ATP5F1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195525.1 | NP_001182454.1 | 8 - 9 | 1390 | 151 | |
| GenBank mRNA | DQ403110.1 | - | 5 - 6 | 755 | 151 | |
More Information
Set Membership: |
|

ATP5F1B
Genomic Map