Assay Details
| Gene Symbol: CS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195515.1 | NP_001182444.1 | 7 - 8 | 943 | 86 | |
| GenBank mRNA | DQ403137.1 | - | 1 - 2 | 118 | 86 | |
More Information
Set Membership: |
|

CS
Genomic Map