Assay Details
| Gene Symbol: RPS27L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003363667.4 | XP_003363715.1 | 377 | 73 | ||
| GenBank mRNA | EF397516.1 | - | 2 - 3 | 216 | 73 | |
More Information
Set Membership: |
|

RPS27L
Genomic Map