Assay Details
| Gene Symbol: DRB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142811.1 | NP_001136283.1 | 5 - 6 | 842 | 94 | |
| XM_023624020.1 | XP_023479788.1 | 598 | 94 | |||
| GenBank mRNA | JQ254085.1 | - | 5 - 6 | 790 | 94 | |
More Information
Set Membership: |
|

DRB
Genomic Map