Assay Details
| Gene Symbol: DRB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142816.1 | NP_001136288.1 | 1 - 2 | 103 | 82 | |
| XM_005603505.3 | XP_005603562.2 | 138 | 82 | |||
| GenBank mRNA | JQ254095.1 | - | 1 - 2 | 171 | 82 | |
| JQ254096.1 | - | 1 - 2 | 171 | 82 | ||
| JQ254097.1 | - | 1 - 2 | 171 | 82 | ||
| JQ254098.1 | - | 1 - 2 | 171 | 82 | ||
More Information
Set Membership: |
|

DRB
Genomic Map