Assay Details
| Gene Symbol: DQA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142814.1 | NP_001136286.1 | 2 | 251 | 92 | |
| XM_005603503.3 | XP_005603560.1 | 323 | 92 | |||
| XM_005603504.3 | XP_005603561.1 | 478 | 92 | |||
| GenBank mRNA | JQ254068.1 | - | 2 | 252 | 92 | |
More Information
Set Membership: |
|

DQA
Genomic Map