Assay Details
| Gene Symbol: TWIST1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146165.1 | NP_001139637.1 | 1 | 451 | 89 | |
| XM_023638777.1 | XP_023494545.1 | 577 | 89 | |||
More Information
Set Membership: |
|

TWIST1
Genomic Map