Assay Details
Gene Symbol: MIR346 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_032788.1 | 1 | 42 | 64 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | glutamate ionotropic receptor delta type subunit 1 | GRID1 | Sense | Chr. 1 - 84873450 - 85543939 on NCBI Build EquCab3.0 | ||
Downstream | coiled-coil serine rich protein 2 | CCSER2 | 1.4mb | Anti-sense | Chr. 1 - 86361053 - 86533449 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
