Assay Details
Gene Symbol: MIR128-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_032966.1 | 1 | 32 | 58 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | R3H domain containing 1 | R3HDM1 | Sense | Chr. 18 - 19434146 - 19615397 on NCBI Build EquCab3.0 | ||
Downstream | UBX domain protein 4 | UBXN4 | 78.1kb | Sense | Chr. 18 - 19637770 - 19673097 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
