Assay Details
| Gene Symbol: IL17A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001143792.1 | NP_001137264.1 | 2 - 3 | 224 | 95 | |
| XM_023624010.1 | XP_023479778.1 | 354 | 95 | |||
| GenBank mRNA | AY014959.1 | - | 1 - 2 | 168 | 95 | |
| EU744563.2 | - | 2 - 3 | 224 | 95 | ||
| KX345375.1 | - | 1 - 2 | 155 | 95 | ||
More Information
Set Membership: |
|

IL17A
Genomic Map