Assay Details
| Gene Symbol: CD1B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163862.1 | NP_001157334.1 | 5 - 6 | 999 | 96 | |
| GenBank mRNA | JL621734.1 | - | 5 - 6 | 1067 | 96 | |
More Information
Set Membership: |
|

CD1B1
Genomic Map