Assay Details
| Gene Symbol: CYCS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164014.1 | NP_001157486.1 | 1 - 2 | 168 | 101 | |
| XM_005609174.3 | XP_005609231.1 | 320 | 101 | |||
More Information
Set Membership: |
|

CYCS
Genomic Map