Assay Details
| Gene Symbol: CD1B2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001246676.1 | NP_001233605.1 | 3 - 4 | 608 | 94 | |
| XM_023640267.1 | XP_023496035.1 | 1533 | 94 | |||
| GenBank mRNA | JF749876.1 | - | 3 - 4 | 608 | 94 | |
More Information
Set Membership: |
|

CD1B2
Genomic Map