Assay Details
| Gene Symbol: HAMP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168327.2 | NP_001161799.1 | 2 - 3 | 194 | 74 | |
| GenBank mRNA | BM780590.1 | - | 2 - 3 | 202 | 74 | |
| BM780771.1 | - | 2 - 3 | 183 | 74 | ||
| GQ253624.1 | - | 2 - 3 | 158 | 74 | ||
More Information
Set Membership: |
|

HAMP
Genomic Map