Assay Details
| Gene Symbol: SELENOH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168387.1 | NP_001161859.1 | 1 - 2 | 315 | 55 | |
| GenBank mRNA | DN504565.1 | - | 1 - 2 | 315 | 55 | |
More Information
Set Membership: |
|

SELENOH
Genomic Map