Assay Details
| Gene Symbol: PEG10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172558.2 | NP_001166029.3 | 1 - 2 | 256 | 86 | |
| NM_001172559.2 | NP_001166030.3 | 1 - 2 | 256 | 86 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001172558.2, NM_001172559.2 transcripts.
Set Membership: |
|

PEG10
Genomic Map